Learn More
Cayman Chemical HIV-1 Integrase Inhibitor 1, Purity: ≥98%, CAS: 729607-74-3, 10 mg
SDP
Catalog No. 502909728
Encompass_Preferred
An inhibitor of HIV-1 integrase DNA strand transfer (IC50 = 14 nM); an inhibitor of SARS-CoV-2 Mpro at 10 µM in a fluorescence-based assay; decreases SARS-CoV-2 replication (EC50 = 0.27 µM) and reduces PFUs in infected Huh7.5 cells in a concentration-dependent manner
Catalog No. 50-290-9728 Supplier Cayman Chemical Supplier No. 3680710 MG
An inhibitor of HIV-1 integrase DNA strand transfer (IC50 = 14 nM); an inhibitor of SARS-CoV-2 Mpro at 10 µM in a fluorescence-based assay; decreases SARS-CoV-2 replication (EC50 = 0.27 µM) and reduces PFUs in infected Huh7.5 cells in a concentration-dependent manner