Learn More
HIMEDIA HIVEG HYDROLYSATE NO. 1 25KG
Catalog No. NC2020563
Encompass_Preferred
HiVeg Hydrolysate No. 1; used for Growth performance at par with Tryptose, enzymatic hydrolysate of protein that can replace tryptose. HIMEDIA mfg#RM030V
Catalog No. NC2020563 Supplier HIMEDIA Supplier No. RM030V25KG
HiVeg Hydrolysate No. 1; used for Growth performance at par with Tryptose, enzymatic hydrolysate of protein that can replace tryptose. HIMEDIA mfg#RM030V