Learn More
HIMEDIA HiVeg Peptone No. 1, Granulated, RMG635V, Microbiology Culture Media Ingredients, Plant Peptone, Animal-Component-Free, Meat Peptone Replacement, Free Flowing, Granules, 25 kg
Catalog No. NC4013795
Encompass_Preferred
HiVeg Peptone No. 1, Granulated is an animal-component-free enzymatic hydrolysate of selected vegetable proteins that directly replaces meat peptone in bacteriological culture media. It delivers the amino acids and peptides needed to cultivate a broad range of micro-organisms while eliminating animal-origin risk, an advantage for regulated and export-sensitive manufacturing. The granulated form flows freely and resists lumping for clean, repeatable media preparation. Store in a cool dry place. Packaged as 25 kg.
Catalog No. NC4013795 Supplier HIMEDIA Supplier No. RMG635V25KG
HiVeg Peptone No. 1, Granulated is an animal-component-free enzymatic hydrolysate of selected vegetable proteins that directly replaces meat peptone in bacteriological culture media. It delivers the amino acids and peptides needed to cultivate a broad range of micro-organisms while eliminating animal-origin risk, an advantage for regulated and export-sensitive manufacturing. The granulated form flows freely and resists lumping for clean, repeatable media preparation. Store in a cool dry place. Packaged as 25 kg.