Learn More
Aviva Systems Biology Corp ENO1 Antibody - middle region (P100955_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100955_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ENO1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ENO1. Alternate names: NNE, PPH, MPB1, ENO1L1ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-53927