Learn More
Selleck Chemical LLC HJC0152 5mg 1420290-99-8
SDP
Supplier:  Selleck Chemical LLC S85615MG
Encompass_Preferred
HJC0152 is a signal transducer and activator of transcription 3 (STAT3) inhibitor with remarkably improved aqueous solubility. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8233