Learn More
Medchemexpress LLC HY-117958 5mg Medchemexpress, HJC0197 CAS:1383539-73-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1179585MG
Encompass_Preferred
Medchemexpress, HY-117958 5mg HJC0197 CAS:1383539-73-8 HJC0197 is a potent Epac1 (exchange protein directly activated by cAMP 1) and Epac2 (IC50=5.9 μM for Epac2) antagonist. HJC0197 selectively blocks cAMP-induced Epac activation. HJC0197 inhibits Epac1-mediated Rap1-GDP exchange activity at 25 μM in the presence of equal concentration of cAMP. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9837