Learn More
Medchemexpress LLC HY-15702 5mg Medchemexpress, HJC0350 CAS:885434-70-8 Purity:>98%
SDP
Catalog No. 502081772
Encompass_Preferred
Medchemexpress, HY-15702 5mg HJC0350 CAS:885434-70-8 HJC0350 is a potent and specific EPAC2 antagonist with an IC50 of 0.3 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1772 Supplier Medchemexpress LLC Supplier No. HY157025MG
Medchemexpress, HY-15702 5mg HJC0350 CAS:885434-70-8 HJC0350 is a potent and specific EPAC2 antagonist with an IC50 of 0.3 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.