Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HJC0416 hydrochloride | 2415263-08-8 | 98.7% | 429.32 g/mol | C18H18Cl2N2O4S | 10MM 1ML
SDP

Supplier:  Medchemexpress LLC HY12352A10MM 1ML

Encompass_Preferred

HJC0416 hydrochloride is a small-molecule inhibitor of STAT3 supplied for laboratory research. It is provided as a 10 mM solution in DMSO (1 mL) and in solid forms, and is reported with CAS 2415263-08-8, molecular weight 429.32 g/mol, formula C18H18Cl2N2O4S, and purity 98.7%.

  • Small-molecule STAT3 inhibitor for cellular assays.
  • Provided as 10 mM solution in DMSO (1 mL) and as solids (5 mg, 10 mg, 25 mg, 50 mg).
  • High reported purity (98.7%).
  • Characterized by CAS 2415263-08-8 and MW 429.32 g/mol.
  • Intended for in vitro research applications targeting STAT3 signaling.

Catalog No. 50-002-38404


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.