Learn More
Medchemexpress LLC HJC0416 hydrochloride | 2415263-08-8 | 98.7% | 429.32 g/mol | C18H18Cl2N2O4S | 25MG
SDP
Catalog No. 5000237715
Encompass_Preferred
HJC0416 hydrochloride is the hydrochloride salt of HJC0416, a potent, orally active small-molecule STAT3 inhibitor used in preclinical anticancer research. Supplied as a white to off-white solid with high chemical purity, it is intended for in vitro and in vivo studies assessing STAT3 pathway modulation.
  • Potent STAT3 inhibition in preclinical models.
  • Oral activity demonstrated in animal studies.
  • High purity suitable for research applications.
  • Available in small research pack sizes, including 25 mg.
  • Store protected from light and under inert gas at recommended temperatures.
Catalog No. 50-002-37715 Supplier Medchemexpress LLC Supplier No. HY12352A25MG
HJC0416 hydrochloride is the hydrochloride salt of HJC0416, a potent, orally active small-molecule STAT3 inhibitor used in preclinical anticancer research. Supplied as a white to off-white solid with high chemical purity, it is intended for in vitro and in vivo studies assessing STAT3 pathway modulation.
  • Potent STAT3 inhibition in preclinical models.
  • Oral activity demonstrated in animal studies.
  • High purity suitable for research applications.
  • Available in small research pack sizes, including 25 mg.
  • Store protected from light and under inert gas at recommended temperatures.