Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HK1/Hexokinase-1 protein, human (His) | >95.0% | 100UG
SDP

Catalog No. 5000174445
Encompass_Preferred

HK1/Hexokinase-1 Protein, Human (His) is a recombinant, His-tagged fragment of human hexokinase-1 (residues E13-A475) produced in E. coli and supplied lyophilized for research use. The product is >95% pure by reducing SDS-PAGE, has an approximate molecular weight of 55.9 kDa, and is formulated with trehalose for stability. Storage and recommended reconstitution guidance are provided for optimal activity and long-term storage.

  • Recombinant fragment corresponding to human HK1 (E13-A475).
  • N-terminal 6xHis tag for purification and detection.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Stable stored at -20°C; aliquot and store at -80°C for long term.
  • Suitable for enzymatic assays, antibody validation, and biochemical studies.

Catalog No. 50-001-74445 Supplier Medchemexpress LLC Supplier No. HYP700580100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HK1/Hexokinase-1 Protein, Human (His) is a recombinant, His-tagged fragment of human hexokinase-1 (residues E13-A475) produced in E. coli and supplied lyophilized for research use. The product is >95% pure by reducing SDS-PAGE, has an approximate molecular weight of 55.9 kDa, and is formulated with trehalose for stability. Storage and recommended reconstitution guidance are provided for optimal activity and long-term storage.

  • Recombinant fragment corresponding to human HK1 (E13-A475).
  • N-terminal 6xHis tag for purification and detection.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Stable stored at -20°C; aliquot and store at -80°C for long term.
  • Suitable for enzymatic assays, antibody validation, and biochemical studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.