Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexokinase-1 protein, human (His-tagged) | >95.0% | 10UG
SDP

Catalog No. 5000176941
Encompass_Preferred

Recombinant human Hexokinase-1 (HK1) fragment (E13-A475) with an N-6xHis tag, expressed in E. coli and supplied as a lyophilized, high-purity protein for enzymatic assays, binding studies, and structural analysis. The product is provided in multiple pack sizes and includes formulation components to enhance stability during storage and handling.

  • Recombinant human fragment E13-A475.
  • N-6xHis tag for affinity purification.
  • Expressed in E. coli for non-glycosylated protein.
  • >95.0% purity by reducing SDS-PAGE.
  • Molecular weight 55.9 kDa (approx.).
  • Lyophilized formulation with trehalose for stability.
  • Store at -20°C; freeze aliquots at -20°C or -80°C for long-term.

Catalog No. 50-001-76941 Supplier Medchemexpress LLC Supplier No. HYP70058010UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Hexokinase-1 (HK1) fragment (E13-A475) with an N-6xHis tag, expressed in E. coli and supplied as a lyophilized, high-purity protein for enzymatic assays, binding studies, and structural analysis. The product is provided in multiple pack sizes and includes formulation components to enhance stability during storage and handling.

  • Recombinant human fragment E13-A475.
  • N-6xHis tag for affinity purification.
  • Expressed in E. coli for non-glycosylated protein.
  • >95.0% purity by reducing SDS-PAGE.
  • Molecular weight 55.9 kDa (approx.).
  • Lyophilized formulation with trehalose for stability.
  • Store at -20°C; freeze aliquots at -20°C or -80°C for long-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.