Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant human hexokinase-1 (His tag) | >95.0% | 50UG
SDP

Catalog No. 5000175485
Encompass_Preferred

Recombinant human hexokinase-1 (HK1) is a His-tagged protein expressed in E. coli and supplied lyophilized. The enzyme catalyzes phosphorylation of hexose sugars to their 6-phosphate derivatives and is provided for enzymatic assays, structural studies, and antibody production applications.

  • N-terminal His tag for purification and detection.
  • Expressed in E. coli and lyophilized for stability.
  • Supplied as 50 μg amount.
  • Molecular weight ~55.9 kDa.
  • Purity >95% by reducing SDS-PAGE.
  • Low endotoxin level <1 EU/μg (LAL).
  • Formulated in Tris/PBS-based buffer with 6% trehalose.
  • Store at -20°C; aliquot and freeze at -80°C for long-term storage.

Catalog No. 50-001-75485 Supplier Medchemexpress LLC Supplier No. HYP70058050UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human hexokinase-1 (HK1) is a His-tagged protein expressed in E. coli and supplied lyophilized. The enzyme catalyzes phosphorylation of hexose sugars to their 6-phosphate derivatives and is provided for enzymatic assays, structural studies, and antibody production applications.

  • N-terminal His tag for purification and detection.
  • Expressed in E. coli and lyophilized for stability.
  • Supplied as 50 μg amount.
  • Molecular weight ~55.9 kDa.
  • Purity >95% by reducing SDS-PAGE.
  • Low endotoxin level <1 EU/μg (LAL).
  • Formulated in Tris/PBS-based buffer with 6% trehalose.
  • Store at -20°C; aliquot and freeze at -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.