Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HKGreen-4I | 1448821-82-6 | 99.9% | 666.63 | 100 MG
SDP

Catalog No. 5000428174
Encompass_Preferred

HKGreen-4I is a highly sensitive green fluorescent probe designed for the specific detection of ONOO- (peroxynitrite) in living cells. It has a maximum excitation wavelength of 520 nm and an emission wavelength of 543 nm. This product is for research use only and not sold to patients.

  • Highly sensitive green fluorescent probe
  • Specific detection of ONOO- in living cells
  • Maximum excitation wavelength: 520 nm
  • Emission wavelength: 543 nm

Catalog No. 50-004-28174 Supplier Medchemexpress LLC Supplier No. HYD1148100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

HKGreen-4I is a highly sensitive green fluorescent probe designed for the specific detection of ONOO- (peroxynitrite) in living cells. It has a maximum excitation wavelength of 520 nm and an emission wavelength of 543 nm. This product is for research use only and not sold to patients.

  • Highly sensitive green fluorescent probe
  • Specific detection of ONOO- in living cells
  • Maximum excitation wavelength: 520 nm
  • Emission wavelength: 543 nm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.