Learn More
TARGETMOL CHEMICALS INC HKI-357 5MG
SDP
Catalog No. 502880664
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. HKI-357 is an irreversible dual inhibitor of EGFR and ERBB2 (IC50s 34 nM and 33 nM), effectively suppressing EGFR autophosphorylation (at Y1068), as well as AKT and MAPK phosphorylation. Purity 98.31%
Catalog No. 50-288-0664 Supplier TARGETMOL CHEMICALS INC Supplier No. T115695MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. HKI-357 is an irreversible dual inhibitor of EGFR and ERBB2 (IC50s 34 nM and 33 nM), effectively suppressing EGFR autophosphorylation (at Y1068), as well as AKT and MAPK phosphorylation. Purity 98.31%