Learn More
Medchemexpress LLC HKI-357 | 848133-17-5 | MFCD26792555 | 99.2% | 574.05 g/mol | C31H29ClFN5O3 | 100 MG
SDP
Catalog No. 5000247742
Encompass_Preferred
HKI-357 is a small-molecule, irreversible dual inhibitor of EGFR and ERBB2 with low-nanomolar potency. It inhibits EGFR (IC50 34 nM) and ERBB2 (IC50 33 nM), suppresses EGFR autophosphorylation at Y1068, and reduces AKT and MAPK phosphorylation. HKI-357 is used in biochemical assays and cell-based studies to probe EGFR/ERBB2 signaling and evaluate inhibitor activity in preclinical cancer research.
  • Irreversible dual inhibitor of EGFR and ERBB2 with IC50s of 34 nM and 33 nM.
  • Suppresses EGFR autophosphorylation (Y1068) and downstream AKT and MAPK phosphorylation.
  • High purity (99.2% by HPLC) suitable for research applications.
  • Available in milligram quantities for dose-ranging and assay setup.
  • Suited for biochemical kinase assays and cell signaling studies.
Catalog No. 50-002-47742 Supplier Medchemexpress LLC Supplier No. HY103443100MG
HKI-357 is a small-molecule, irreversible dual inhibitor of EGFR and ERBB2 with low-nanomolar potency. It inhibits EGFR (IC50 34 nM) and ERBB2 (IC50 33 nM), suppresses EGFR autophosphorylation at Y1068, and reduces AKT and MAPK phosphorylation. HKI-357 is used in biochemical assays and cell-based studies to probe EGFR/ERBB2 signaling and evaluate inhibitor activity in preclinical cancer research.
  • Irreversible dual inhibitor of EGFR and ERBB2 with IC50s of 34 nM and 33 nM.
  • Suppresses EGFR autophosphorylation (Y1068) and downstream AKT and MAPK phosphorylation.
  • High purity (99.2% by HPLC) suitable for research applications.
  • Available in milligram quantities for dose-ranging and assay setup.
  • Suited for biochemical kinase assays and cell signaling studies.