Learn More
Aviva Systems Biology Corp HKR1 Antibody - N-terminal region (ARP40040_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40040_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HKR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HKR1. Alternate names: ZNF875ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-57273