Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HKSOX-1m (5/6-mixture) hydrobromide | 1786411-19-5 | 99.6% | 1258.72 | C49H37Br2F10N2O10PS2 | 10 MG
SDP

Catalog No. 5000297178
Encompass_Preferred

HKSOX-1m (5/6-mixture) hydrobromide is a mitochondria-targeted O2 (superoxide) fluorescent probe that fluoresces green (Ex/Em 509/534 nm). It enables sensitive imaging of basal and Antimycin A-stimulated mitochondrial superoxide in live cells, with high selectivity across a broad pH range and recommended sealed storage to protect from moisture and light.

  • Mitochondria-targeted O2 (superoxide) fluorescent probe.
  • Excitation/emission: 509/534 nm (green fluorescence).
  • High reported purity: 99.55%.
  • Molecular weight: 1258.72 g/mol.
  • Appearance: light yellow to orange solid.
  • Storage: sealed, away from moisture and light; in solvent -80 °C (6 months), -20 °C (1 month).

Catalog No. 50-002-97178 Supplier Medchemexpress LLC Supplier No. HYD1156A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HKSOX-1m (5/6-mixture) hydrobromide is a mitochondria-targeted O2 (superoxide) fluorescent probe that fluoresces green (Ex/Em 509/534 nm). It enables sensitive imaging of basal and Antimycin A-stimulated mitochondrial superoxide in live cells, with high selectivity across a broad pH range and recommended sealed storage to protect from moisture and light.

  • Mitochondria-targeted O2 (superoxide) fluorescent probe.
  • Excitation/emission: 509/534 nm (green fluorescence).
  • High reported purity: 99.55%.
  • Molecular weight: 1258.72 g/mol.
  • Appearance: light yellow to orange solid.
  • Storage: sealed, away from moisture and light; in solvent -80 °C (6 months), -20 °C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.