Learn More
Medchemexpress LLC HKSOX-1r (5/6-mixture) | 98.4% | 50 MG
SDP
Catalog No. 5000340361
Encompass_Preferred
HKSOX-1r (5/6-mixture) is a fluorescent probe designed for imaging and detecting endogenous superoxide in live cells and in vivo. This probe demonstrates excellent selectivity and sensitivity towards the superoxide anion radical, making it suitable for various research applications.
  • Fluorescent probe for imaging and detecting endogenous superoxide
  • Used in live cells and in vivo
  • Exhibits excellent selectivity and sensitivity towards superoxide anion radical
  • Detects mitochondrial respiratory inhibitor-induced O2•- formation in various cell lines
  • Includes protocols for in vitro working solution preparation
  • Proven to detect fluorescence distribution in zebrafish embryos
Catalog No. 50-003-40361 Supplier Medchemexpress LLC Supplier No. HY13001750MG
HKSOX-1r (5/6-mixture) is a fluorescent probe designed for imaging and detecting endogenous superoxide in live cells and in vivo. This probe demonstrates excellent selectivity and sensitivity towards the superoxide anion radical, making it suitable for various research applications.
  • Fluorescent probe for imaging and detecting endogenous superoxide
  • Used in live cells and in vivo
  • Exhibits excellent selectivity and sensitivity towards superoxide anion radical
  • Detects mitochondrial respiratory inhibitor-induced O2•- formation in various cell lines
  • Includes protocols for in vitro working solution preparation
  • Proven to detect fluorescence distribution in zebrafish embryos