Learn More
Eurofins Discoverx PrecisION hKv7.2/hKv7.3 Recombinant Stable Cell Line, 2 vials. (CYL3059)
Supplier:  Eurofins Discoverx CYL3059
Encompass
The PrecisION hKv7.2/hKv7.3 Recombinant Stable Cell Line is a stable, engineered cell line for use in studying Kv7.2/Kv7.3 ion channels, including drug candidates which affect channel function. This cell line enables assessment of ion channel function via detection of electrophysiological signals. The included cell line overexpresses Kv7.2/Kv7.3 ion channels. Upon ligand/agonist induced channel activation, response can be detected via automated or manual electrophysiological techniques. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/ion-channels.
Catalog No. 50-200-5092
Add to Cart
Request a Quote