Learn More
Medchemexpress LLC HKYellow-AM (6/12-mixture) | 1448821-89-3 | 99.4% | 1 MG
SDP
Catalog No. 5000340831
Encompass_Preferred
HKYellow-AM (6/12-mixture) is a yellow fluorescent probe designed to detect ONOO- in living cells and tissues. It offers high selectivity and sensitivity without exhibiting cytotoxicity, making it suitable for research applications.
  • Yellow fluorescent probe
  • Detects ONOO- in living cells and tissues
  • High selectivity
  • High sensitivity
  • No cytotoxicity
Catalog No. 50-003-40831 Supplier Medchemexpress LLC Supplier No. HY1300131MG
HKYellow-AM (6/12-mixture) is a yellow fluorescent probe designed to detect ONOO- in living cells and tissues. It offers high selectivity and sensitivity without exhibiting cytotoxicity, making it suitable for research applications.
  • Yellow fluorescent probe
  • Detects ONOO- in living cells and tissues
  • High selectivity
  • High sensitivity
  • No cytotoxicity