Learn More
ATCC HL-60, Promyelocytic Leukemia, Frozen
Supplier:  ATCC CCL240/DEL
Encompass
HL-60 cells spontaneously differentiate and differentiation can be stimulated by butyrate, hypoxanthine, phorbol myristic acid (PMA, TPA), dimethylsulfoxide (DMSO, 1% to 1.5%), actinomycin D, and retinoic acid. The cells exhibit phagocytic activity and responsiveness to chemotactic stimuli. The line is positive for myc oncogene expression.
Catalog No. 50-188-353FP