Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rotronic Instrument Corp HYGROLOG HL-20D-SET1 Data Logger
SDP

Catalog No. 18700110 Shop All Rotronic Instrument Corp Products
Encompass_Preferred

The compact data logger for humidity and temperature measurement offers high precision and reliability at a reasonable price. The HL-20 series is easy to use and deployable in a wide range of applications. Thanks to its integrated batteries, the HL-20 works completely independently and offers its users maximum flexibility.

Applications: Warehouses, factory halls, museums, office buildings, clean rooms, shipping, libraries, test facilities

Features:

  • Range of application: -10…60 °C, 0…100 %rh
  • 20’000 data point memory
  • Freely selectable logging interval, 5 s…1 h
  • Integrated clock enables timestamp for every measurement
  • Adjusted at 10, 35, 80 %rh and 23 °C
  • Programmable alarms• Battery operation
  • Interface (UART) for connection to PC
  • No software

Catalog No. 18-700-110 Supplier Rotronic Instrument Corp Supplier No. HL20DSET1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The compact data logger for humidity and temperature measurement offers high precision and reliability at a reasonable price. The HL-20 series is easy to use and deployable in a wide range of applications. Thanks to its integrated batteries, the HL-20 works completely independently and offers its users maximum flexibility.

Applications: Warehouses, factory halls, museums, office buildings, clean rooms, shipping, libraries, test facilities

Features:

  • Range of application: -10…60 °C, 0…100 %rh
  • 20’000 data point memory
  • Freely selectable logging interval, 5 s…1 h
  • Integrated clock enables timestamp for every measurement
  • Adjusted at 10, 35, 80 %rh and 23 °C
  • Programmable alarms• Battery operation
  • Interface (UART) for connection to PC
  • No software

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.