Learn More
Enzo Life Sciences HL60 Cell Extract (non-induced) (200 µl)
SDP
Supplier:  Enzo Life Sciences BMLSW1010200
Encompass_Preferred
Immunoblotting standard. Whole-cell extract of human HL-60 leukemia cells. Supplied in SDS-PAGE sample buffer, ready-to-use. Applications: WB control for PARP antibodies, to detect intact 116 kDa PARP. Recommended gel load is 20 µl per lane (~75,000 cells). Formulation: Cell extract in SDS-PAGE sample buffer (62.5 mM Tris/HCl, pH 6.8, containing 6 M urea, 10% glycerol, 2% SDS, 0.00125% bromophenol blue and 5% β-mercaptoethanol). The sample is ready to use for SDS-PAGE. Long Term Storage: -20°C.
Catalog No. 50-201-2194