Learn More
ATCC HL-60; Promyelocytic Leukemia; Genomic DNA, Human
Catalog No. 502384266
Encompass
High molecular weight genomic DNA isolated from the HL-60 human acute promyelocytic leukemia cell line (ATCC CCL-240). This cell line is positive for myc oncogene expression.
Catalog No. 50-238-4266 Supplier ATCC Supplier No. CCL240D
High molecular weight genomic DNA isolated from the HL-60 human acute promyelocytic leukemia cell line (ATCC CCL-240). This cell line is positive for myc oncogene expression.