Learn More
ATCC HL-60; Promyelocytic Leukemia; Human (Homo sapiens)
Catalog No. 502384462
Encompass
HL-60 cells are promyeoloblasts isolated from the peripheral blood by leukopheresis from a 36-year-old,White, female with acute promyelocytic leukemia. This cell line can be used in immune disorder and immunology research.
Catalog No. 50-238-4462 Supplier ATCC Supplier No. CCL240
 Restricted Item
Add to Cart
Add to Cart
HL-60 cells are promyeoloblasts isolated from the peripheral blood by leukopheresis from a 36-year-old,White, female with acute promyelocytic leukemia. This cell line can be used in immune disorder and immunology research.