Learn More
ACROBiosystems Human HLA-A*02:01&B2M&WT-1 (RMFPNAPYL) Tetramer Protein (MALS verified)
Catalog No. 502331854
Encompass_Preferred
Human HLA-A*02:01&B2M&WT-1 (RMFPNAPYL) Tetramer Protein (HLW-H52E6) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*02:01) & Ile 21 - Met 119 (B2M) & RMFPNAPYL peptide (Accession # AAA59606.1 (HLA-A*02:01) & P61769 (B2M) & RMFPNAPYL).
Catalog No. 50-233-1854 Supplier ACROBiosystems Supplier No. HLWH52E625UG
Human HLA-A*02:01&B2M&WT-1 (RMFPNAPYL) Tetramer Protein (HLW-H52E6) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*02:01) & Ile 21 - Met 119 (B2M) & RMFPNAPYL peptide (Accession # AAA59606.1 (HLA-A*02:01) & P61769 (B2M) & RMFPNAPYL).