Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HLA-A*02:01&B2M&PRAME (SLLQHLIGL) monomer protein, human | >95.0% | 50UG
SDP

Catalog No. 5000176727
Encompass_Preferred

Recombinant human HLA-A*02:01 monomer complexed with beta-2 microglobulin and the PRAME-derived peptide SLLQHLIGL. Produced in HEK293 cells and supplied with C-terminal Avi and 8xHis tags, this high-purity monomer is intended for antigen presentation research, epitope mapping, and T cell assays.

  • Monomeric HLA-A*02:01 complex with PRAME peptide SLLQHLIGL for epitope-specific assays.
  • Produced in HEK293 cells for native folding and appropriate post-translational modifications.
  • C-terminal Avi tag for biotinylation and C-terminal 8xHis tag for purification and detection.
  • Purity >95.0% as determined by reducing SDS-PAGE.
  • Available in small, pre-measured sizes suitable for T cell assays and ELISPOT.
  • Stable when stored at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.

Catalog No. 50-001-76727 Supplier Medchemexpress LLC Supplier No. HYP70109050UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human HLA-A*02:01 monomer complexed with beta-2 microglobulin and the PRAME-derived peptide SLLQHLIGL. Produced in HEK293 cells and supplied with C-terminal Avi and 8xHis tags, this high-purity monomer is intended for antigen presentation research, epitope mapping, and T cell assays.

  • Monomeric HLA-A*02:01 complex with PRAME peptide SLLQHLIGL for epitope-specific assays.
  • Produced in HEK293 cells for native folding and appropriate post-translational modifications.
  • C-terminal Avi tag for biotinylation and C-terminal 8xHis tag for purification and detection.
  • Purity >95.0% as determined by reducing SDS-PAGE.
  • Available in small, pre-measured sizes suitable for T cell assays and ELISPOT.
  • Stable when stored at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.