Learn More
Immudex HLA-A 0201 SLLMWITQC PE, 150 tests
SDP
Catalog No. NC1467424
Encompass
The Immudex MHC Dextramer reagent is specifically designed for the precise detection of antigen-specific T cells through flow cytometry. This reagent features a dextran polymer backbone adorned with HLA-A*02:01-peptide complexes, particularly the SLLMWITQC peptide, conjugated with the PE fluorochrome for enhanced visualization. It is intended solely for research applications, providing high avidity binding to identify low-affinity T-cell populations effectively.
  • High avidity binding enhances detection of low-affinity T cells and reduces background staining.
  • Sensitive detection enables flow cytometric identification of rare CD4+ and CD8+ T cell populations.
  • Suitable for diverse research fields, including immune-oncology and autoimmunity.
  • Exclusively for research use, not for diagnostic or therapeutic applications.
  • Manufactured under cGMP standards for consistency and reliability.
Catalog No. NC1467424 Supplier Immudex Supplier No. WB02696 PE 150
The Immudex MHC Dextramer reagent is specifically designed for the precise detection of antigen-specific T cells through flow cytometry. This reagent features a dextran polymer backbone adorned with HLA-A*02:01-peptide complexes, particularly the SLLMWITQC peptide, conjugated with the PE fluorochrome for enhanced visualization. It is intended solely for research applications, providing high avidity binding to identify low-affinity T-cell populations effectively.
  • High avidity binding enhances detection of low-affinity T cells and reduces background staining.
  • Sensitive detection enables flow cytometric identification of rare CD4+ and CD8+ T cell populations.
  • Suitable for diverse research fields, including immune-oncology and autoimmunity.
  • Exclusively for research use, not for diagnostic or therapeutic applications.
  • Manufactured under cGMP standards for consistency and reliability.