Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3105MSM4XXP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM417. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: THP-1, Jurkat cells. Tonsil or lymph node. Cellular Staining: Cell Surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Normal human peripheral blood lymphocytes of phenotype A1, Aw32, B7, B37, Cw-, Cw-, DR2, DRw10. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. Chromosome: 6q21.3. Gene Symbol: HLA-A. Gene ID: 3105. Ensemble: ENSG00000223980. Uniprot: XP_016885777.
Catalog No. 50-188-4770