Learn More
KACTUSBIO INC Human HLA-A*11:01&B2M&KRAS G12R (VVVGARGVGK) Tetramer Protein (MHC-HM440T) . 1EA=100UG.
Supplier:  KACTUSBIO INC MHCHM440T100UG
Encompass
Recombinant Human HLA-A*11:01&B2M&KRAS G12R (VVVGARGVGK) Tetramer Protein is expressed from HEK293 with His tag and Avi tag at the C-Terminus, tetramer is assembled by biotinylated monomer and streptavidin.It contains Gly25-Thr305(HLA-A*11:01), Ile21-Met119(B2M) and VVVGARGVGK peptide.
Catalog No. NC2956800