Learn More
ACROBiosystems Biotinylated Human HLA-A*0201 GP100 (YLEPGPVTA) Complex Protein (HLG-H82E6) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*0201) & YLEPGPVTA & Ile 21 - Met 119 (B2M) (Accession # A0A140T913 (HLA-A*0201) &
Catalog No. 502254089
Encompass_Preferred
Glycoprotein 100, is also well known as gp100 and Melanocyte protein PMEL. This antigen is a melanocyte-specific type I transmembrane glycoprotein and composed of 661 amino acids. Glycoprotein 100 is related to the tumor regression in metastatic melanoma patients after adoptive therapy, by which its epitopes being recognized by tumor lymphocytes and T-cells. The gp100280–288(YLEPGPVTA) was originally shown to be recognized by HLA-A*0201 tumor-infiltrating lymphocytes from melanoma patients, and therefore it is widely been studied in TCR -T studies.
This item is not returnable. View return policy
Catalog No. 50-225-4089 Supplier ACROBiosystems Supplier No. HLGH82E625UG
Glycoprotein 100, is also well known as gp100 and Melanocyte protein PMEL. This antigen is a melanocyte-specific type I transmembrane glycoprotein and composed of 661 amino acids. Glycoprotein 100 is related to the tumor regression in metastatic melanoma patients after adoptive therapy, by which its epitopes being recognized by tumor lymphocytes and T-cells. The gp100280–288(YLEPGPVTA) was originally shown to be recognized by HLA-A*0201 tumor-infiltrating lymphocytes from melanoma patients, and therefore it is widely been studied in TCR -T studies.