Learn More
ACROBiosystems Biotinylated Human HLA-A*0201 WT-1 (RMFPNAPYL) Complex Protein (HLW-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*0201) & RMFPNAPYL & Ile 21 - Met 119 (B2M) (Accession # A0A140T913 (HLA-A*0201) &
Catalog No. 502254088
Encompass_Preferred
Wilms tumor gene 1 (WT1),is an attractive target antigen for leukemia and solid cancer. WT1-specific adoptive immunotherapy has developed for tumor treatment in recent years. WT1 has been proved wildly expressed in breast, colon and ovarian cancer. It participates in cell growth, differentiation and apoptosis regulation. The WT1127-134 (RMFPNAPYL) was shown to be recognized by HLA-A*0201 tumor-infiltrating lymphocytes from melanoma patients, and therefore it is widely been studied in TCR-T studies.
This item is not returnable. View return policy
Catalog No. 50-225-4088 Supplier ACROBiosystems Supplier No. HLWH82E5200UG
Wilms tumor gene 1 (WT1),is an attractive target antigen for leukemia and solid cancer. WT1-specific adoptive immunotherapy has developed for tumor treatment in recent years. WT1 has been proved wildly expressed in breast, colon and ovarian cancer. It participates in cell growth, differentiation and apoptosis regulation. The WT1127-134 (RMFPNAPYL) was shown to be recognized by HLA-A*0201 tumor-infiltrating lymphocytes from melanoma patients, and therefore it is widely been studied in TCR-T studies.