Learn More
Genscript Corporation HLA-A0201B2MHIV Gag SLYNT
Catalog No. 5000056900
Encompass
Gag is the HIV structural precursor protein which is cleaved by viral protease to produce mature infectious viruses Gag is a polyprotein composed of MA (matrix) CA (capsid) SP1 NC (nucleocapsid) SP2 and p6 domains The immunodominant HLA-A2-restricted Gag epitope SLYNTVATL (SL9) is considered to be a poor immunogen because reactivity to it is rare in acute infection despite its paradoxical dominance in patients with chronic infection
Catalog No. 50-000-56900 Supplier Genscript Corporation Supplier No. Z06562100
Gag is the HIV structural precursor protein which is cleaved by viral protease to produce mature infectious viruses Gag is a polyprotein composed of MA (matrix) CA (capsid) SP1 NC (nucleocapsid) SP2 and p6 domains The immunodominant HLA-A2-restricted Gag epitope SLYNTVATL (SL9) is considered to be a poor immunogen because reactivity to it is rare in acute infection despite its paradoxical dominance in patients with chronic infection