Learn More
Aviva Systems Biology Corp HLA-C Antibody - C-terminal region (ARP63059_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63059_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HLA-C. It was validated on Western Blot. . Alternate names: D6S204, FLJ27082, HLA-Cw, HLA-Cw12, HLA-JY3, HLC-C, PSORS1ApplicationWB. Species reactivity: Human, Pig, Rabbit
Catalog No. 50-287-858