Learn More
Aviva Systems Biology Corp HLA-DMB Antibody - N-terminal region (ARP63380_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63380_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HLA-DMB. It was validated on Western Blot. . Alternate names: D6S221E, RING7ApplicationWB, IHC. Species reactivity: Human, Rat
Catalog No. 50-287-952