Learn More
Aviva Systems Biology Corp HLA-DPA1 Antibody - middle region (ARP50199_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50199_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HLA-DPA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HLA-DPA1. Alternate names: HLA-DP1A, HLADP, HLASB, PLT1, DP(W3), DP(W4)ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-60281