Learn More
Aviva Systems Biology Corp HLA-DQA2 Antibody - N-terminal region (ARP49464_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49464_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HLA-DQA2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human HLA-DQA2. Alternate names: DQ alpha, HLA-DXA, DX-ALPHAApplicationWB. Species reactivity: Cow, Goat, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-285-426