Learn More
Biotium HLA-DRB (MHC II)(Bra30), CF594 conjugate, 0.1mg/mL
Supplier:  Biotium BNC940407500
Encompass_Preferred
This MAb reacts with the HLA-DRB3 antigen, a member of MHC class II molecules. It does not cross react with HLA-DP and HLA-DQ. HLA-DR is a heterodimeric cell surface glycoprotein comprised of a 36 kDa alpha (heavy) chain and a 28 kDa beta (light) chain. It is expressed on B-cells, activated T-cells, monocytes/macrophages, dendritic cells and other non-professional APCs. In conjunction with the CD3/TCR complex and CD4 molecules, HLA-DR is critical for efficient peptide presentation to CD4 T cells. HLA-DR antigens also occur on a variety of epithelial cells and their corresponding neoplastic counterparts. Primary antibodies are available purified, or with a selection of fluorescent CF Dyes and other labels. CF Dyes offer exceptional brightness and photostability. Note: Conjugates of blue fluorescent dyes like CF405S and CF405M are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than
Catalog No. 50-252-3120