Learn More
Aviva Systems Biology Corp HLA-E Antibody - C-terminal region (ARP62847_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62847_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HLA-E. It was validated on Western Blot. . Alternate names: DKFZp686P19218, EA1.2, EA2.1, HLA-6.2, MHC, QA1ApplicationWB. Species reactivity: Human, Mouse, Pig
Catalog No. 50-287-784