Learn More
Enzo Life Sciences HLA-E (human) monoclonal antibody (MEM-E/02) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX805701C100
Encompass_Preferred
Clone: MEM-E/02. Isotype: IgG1. Immunogen: Recombinant human HLA-E denatured heavy chain. Species reactivity: Human. Specificity: Recognizes denatured heavy chain of human HLA-E. Specificity was confirmed on HLA-G/HLA-E Workshop (Victoria 2002). Applications: IHC (PS), WB. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 15mM sodium azide.
Catalog No. 50-201-0059