Learn More
Aviva Systems Biology Corp HLA-F Antibody - N-terminal region (ARP49410_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49410_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HLA-F. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human HLA-F. Alternate names: CDA12, HLA-5.4, HLA-CDA12, HLAFApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-285-414