Learn More
Enzo Life Sciences HLA-G (human) monoclonal antibody (MEM-G/9) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX805700C100
Encompass_Preferred
Clone: MEM-G/9. Isotype: IgG1. Immunogen: Recombinant human HLA-G refolded with β2-microglobulin and peptide. Species reactivity: Human. Specificity: Recognizes native HLA-G on the cell surface as well as soluble HLA-G molecule in its β2-microglobulin associated form. Applications: ELISA, Flow Cytometry, ICC, IHC (FS), IP. Purity: Purified by precipitation. Formulation: Liquid. In PBS, pH 7.4, containing 15mM sodium azide.
Catalog No. 50-201-0057