Learn More
Enzo Life Sciences HLA class II Recombinant monoclonal antibody (F3.3) (Mouse IgG1 kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6390200
Encompass_Preferred
Clone: F3.3. Isotype: IgG1κ. Species reactivity: Human. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3163