Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3115MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: CR3/43. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: Raji cells. Tonsil or lymph node. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Cells from human tonsil. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 36kDa (? chain) and 27kDa (? chain).Chromosome: 6p21.3 (HLA-DP ;HLA-DR). Gene Symbol: HLA-DP ;HLA-DQ ;HLA-DR. Gene ID: 3115. Ensemble: ENSG00000223865. Uniprot: NP_002112.
Catalog No. 50-188-4771