Learn More
Biomatik Corporation Hla-Dqb1 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11907
Encompass_Preferred
Hla-Dqb1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to HLA-DQB1. The Hla-Dqb1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Hla Class Ii Histocompatibility Antigen, Dq Beta 1 Chain Protein (128-175Aa). Applications: ELISA, WB, IHC. Target Synonyms: HLA-DQB1. Accession Number: P01920. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7379