Learn More
Cayman Chemical HLADR MoncLNal ANTIBODYC 100ug
SDP
Catalog No. 502986085
Encompass_Preferred
Immunogen: Human lymphoblastoid cell line (RPMI 8866) • Clone designation: L243 • Host: Mouse • Species Reactivity: (+) Human, canine, and non-human primate HLA-DR • Isotype: IgG2a • Applications: FC, ICC, IF, IHC, IP, and WB
Catalog No. 50-298-6085 Supplier Cayman Chemical Supplier No. 21827100 UG
Immunogen: Human lymphoblastoid cell line (RPMI 8866) • Clone designation: L243 • Host: Mouse • Species Reactivity: (+) Human, canine, and non-human primate HLA-DR • Isotype: IgG2a • Applications: FC, ICC, IF, IHC, IP, and WB