Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3123MSM1XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: SPM289. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: Ramos, Daudi or HuT78 cells. Tonsil or lymph node. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Activated human peripheral blood mononuclear cells. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: ~28kDa (beta chain).Chromosome: 6p21.3. Gene Symbol: HLA-DR. Gene ID: 3123. Ensemble: ENSG00000196126. Uniprot: NP_002115.
Catalog No. 50-188-4774