Learn More
Apexbio Technology LLC HLCL-61 1158279-20-9 100mg
SDP
Catalog No. 5000087677
Encompass_Preferred
HLCL-61 (CAS 1158279-20-9) is a selective inhibitor of protein arginine methyltransferase 5 (PRMT5) an enzyme implicated in the epigenetic regulation of gene expression through symmetric dimethylation of arginine residues on histones and other proteins HLCL-61 shows specificity for PRMT5 exhibiting no inhibitory activity against PRMT1 PRMT4 or other type II PRMT family members In cellular studies HLCL-61 suppresses symmetric dimethylation of histones H3 and H4 leading to reduced viability and enhanced differentiation of acute myeloid leukemia (AML) cell lines and primary blasts HLCL-61 is currently utilized in preclinical research to investigate PRMT5-mediated epigenetic mechanisms in cancer
Catalog No. 50-000-87677 Supplier Apexbio Technology LLC Supplier No. B7806100
HLCL-61 (CAS 1158279-20-9) is a selective inhibitor of protein arginine methyltransferase 5 (PRMT5) an enzyme implicated in the epigenetic regulation of gene expression through symmetric dimethylation of arginine residues on histones and other proteins HLCL-61 shows specificity for PRMT5 exhibiting no inhibitory activity against PRMT1 PRMT4 or other type II PRMT family members In cellular studies HLCL-61 suppresses symmetric dimethylation of histones H3 and H4 leading to reduced viability and enhanced differentiation of acute myeloid leukemia (AML) cell lines and primary blasts HLCL-61 is currently utilized in preclinical research to investigate PRMT5-mediated epigenetic mechanisms in cancer