Learn More
Medchemexpress LLC HY-100025A 10mg Medchemexpress, HLCL-61 (hydrochloride) CAS:1158279-20-9 Purity:>98%
SDP
Catalog No. 501931696
Encompass_Preferred
Medchemexpress, HY-100025A 10mg HLCL-61 (hydrochloride) CAS:1158279-20-9 HLCL-61 hydrochloride is a first-in-class inhibitor of protein arginine methyltransferase 5 (PRMT5) . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1696 Supplier Medchemexpress LLC Supplier No. HY100025A10MG
Medchemexpress, HY-100025A 10mg HLCL-61 (hydrochloride) CAS:1158279-20-9 HLCL-61 hydrochloride is a first-in-class inhibitor of protein arginine methyltransferase 5 (PRMT5) . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.