Learn More
Medchemexpress LLC Hlcl-61 hydrochloride | 1158279-20-9 | 99.9% | 380.91 | C23H25ClN2O | 5 MG
SDP
Catalog No. 5000214992
Encompass_Preferred
HLCL-61 hydrochloride is a first-in-class small-molecule inhibitor of protein arginine methyltransferase 5 (PRMT5) used for research to probe PRMT5-mediated methylation and cellular responses. It reduces cell growth in a dose-dependent manner across multiple leukemia cell lines and inhibits symmetric arginine dimethylation of histones H3 and H4, supporting studies of epigenetic regulation and oncology research.
  • Targets PRMT5 to inhibit arginine methylation.
  • Reduces cell proliferation in leukemia cell lines; IC50s reported by manufacturer.
  • High purity suitable for research applications.
  • Soluble in DMSO (50 mg/mL); insoluble in water.
  • Solid, white to off-white appearance; store sealed away from moisture under recommended conditions.
Catalog No. 50-002-14992 Supplier Medchemexpress LLC Supplier No. HY100025A5MG
HLCL-61 hydrochloride is a first-in-class small-molecule inhibitor of protein arginine methyltransferase 5 (PRMT5) used for research to probe PRMT5-mediated methylation and cellular responses. It reduces cell growth in a dose-dependent manner across multiple leukemia cell lines and inhibits symmetric arginine dimethylation of histones H3 and H4, supporting studies of epigenetic regulation and oncology research.
  • Targets PRMT5 to inhibit arginine methylation.
  • Reduces cell proliferation in leukemia cell lines; IC50s reported by manufacturer.
  • High purity suitable for research applications.
  • Soluble in DMSO (50 mg/mL); insoluble in water.
  • Solid, white to off-white appearance; store sealed away from moisture under recommended conditions.